ENA065085932 Enamine

2-[2-oxo-5-(trifluoromethyl)pyridin-1(2H)-yl]acetohydrazide

  • MDL number MFCD02073537
  • Cas Number : 314768-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160...162
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14758

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14758
Availability details for
Quantity
  • Availability
  • Add to Cart