ENA065085942 Enamine

[5-(2-methyl-1,3-thiazol-4-yl)thien-2-yl]methylamine

  • MDL number MFCD02682054
  • Cas Number : 868238-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...227
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14480

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-14480
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart