ENA065085946 Enamine

ethyl 2-(1,1-dioxo-1lambda6-thiolan-3-yl)-3-oxobutanoate

  • MDL number MFCD00188768
  • Cas Number : 4905-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-80
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14483
Availability details for
Quantity
  • Availability
  • Add to Cart