ENA065085950 Enamine

3-[2-(4-methoxyphenyl)-1H-indol-3-yl]propanoic acid

  • MDL number MFCD06660847
  • Cas Number : 870693-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-117
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14776
Availability details for
Quantity
  • Availability
  • Add to Cart