ENA065085951 Enamine

methyl({[2-(piperidin-1-yl)phenyl]methyl})amine

  • MDL number MFCD07348553
  • Cas Number : 871217-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14777
Availability details for
Quantity
  • Availability
  • Add to Cart