ENA065085955 Enamine

1-[(dimethyl-1,2-oxazol-4-yl)sulfonyl]piperidine-4-carboxylic acid

  • MDL number MFCD07343993
  • Cas Number : 697258-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14486
Availability details for
Quantity
  • Availability
  • Add to Cart