ENA065085957 Enamine

4-isobutyl-1,5-dioxo-2,3,4,5-tetrahydropyrrolo[1,2-a]quinazoline-3a(1H)-carboxylic acid

  • MDL number MFCD07339178
  • Cas Number : 863669-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14250

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14250
Availability details for
Quantity
  • Availability
  • Add to Cart