ENA065085959 Enamine

(1-benzyl-3,5-dimethyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD07346343
  • Cas Number : 956567-89-8
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14785
Availability details for
Quantity
  • Availability
  • Add to Cart