ENA065085962 Enamine

N-[4-(2-amino-1,3-thiazol-4-yl)phenyl]methanesulfonamide

  • MDL number MFCD01627908
  • Cas Number : 476338-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 196-197
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14788
Availability details for
Quantity
  • Availability
  • Add to Cart