ENA065085972 Enamine

2-[3-(ethoxycarbonyl)-2,5-dimethyl-1H-pyrrol-1-yl]acetic acid

  • MDL number MFCD07339357
  • Cas Number : 679797-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-52
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14332
Availability details for
Quantity
  • Availability
  • Add to Cart