ENA065085975 Enamine

4-chloro-5-hydrazinyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD01848851
  • Cas Number : 6959-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14496
Availability details for
Quantity
  • Availability
  • Add to Cart