ENA065085976 Enamine

4-[4-(difluoromethoxy)phenyl]-5-methyl-1,3-thiazol-2-amine

  • MDL number MFCD04837239
  • Cas Number : 794578-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-108
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14253
Availability details for
Quantity
  • Availability
  • Add to Cart