ENA065085979 Enamine

2,2-difluoro-1,3-benzodioxol-5-amine

  • MDL number MFCD00190144
  • Cas Number : 1544-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14335

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-14335
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart