ENA065085984 Enamine

4-[(4-phenyl-1,3-thiazol-2-yl)amino]butanoic acid

  • MDL number MFCD05233629
  • Cas Number : 869634-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14500
Availability details for
Quantity
  • Availability
  • Add to Cart