ENA065085987 Enamine

3-(1,3-benzothiazol-2-yl)-2,3-dihydroxypropanoic acid

  • MDL number MFCD02699535
  • Cas Number : 326021-35-6
  • Molecular Weight : 239.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14503
Availability details for
Quantity
  • Availability
  • Add to Cart