ENA065085992 Enamine

3-methyl-5-(methylamino)-1,2-thiazole-4-carboxylic acid

  • MDL number MFCD07348567
  • Cas Number : 871217-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14818
Availability details for
Quantity
  • Availability
  • Add to Cart