ENA065085997 Enamine

3-(3,5-dimethoxyphenyl)propanoic acid

  • MDL number MFCD03425685
  • Cas Number : 717-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 54...56
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14823

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-14823
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart