ENA065086003 Enamine

4-chloro-2-(chloromethyl)-1-methoxybenzene

  • MDL number MFCD02182262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44...46
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14511

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14511
Availability details for
Quantity
  • Availability
  • Add to Cart