ENA065086008 Enamine

3-(4-chlorophenyl)-3-[(phenylsulfonyl)amino]propanoic acid

  • MDL number MFCD03856033
  • Cas Number : 613656-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155...157
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14340
Availability details for
Quantity
  • Availability
  • Add to Cart