ENA065086015 Enamine

5-(4-methylphenyl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD00461061
  • Cas Number : 51884-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229-230
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14520
Availability details for
Quantity
  • Availability
  • Add to Cart