ENA065086019 Enamine

2-[(3,5-dimethyl-1-phenyl-1H-pyrazol-4-yl)sulfanyl]acetic acid

  • MDL number MFCD07339183
  • Cas Number : 93350-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-122
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14259
Availability details for
Quantity
  • Availability
  • Add to Cart