ENA065086031 Enamine

N-(3,4-dimethylphenyl)urea

  • MDL number MFCD00025426
  • Cas Number : 114-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14857

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14857
Availability details for
Quantity
  • Availability
  • Add to Cart