ENA065086032 Enamine

N-(2,3-dimethylphenyl)urea

  • MDL number MFCD00025408
  • Cas Number : 13114-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205...207
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14858

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14858
Availability details for
Quantity
  • Availability
  • Add to Cart