ENA065086034 Enamine

N,N-diethyl-4-[(methylamino)methyl]aniline

  • MDL number MFCD07348593
  • Cas Number : 863646-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14860
Availability details for
Quantity
  • Availability
  • Add to Cart