ENA065086037 Enamine

4,6-dimethyl-2-sulfanylpyridine-3-carboxylic acid

  • MDL number MFCD06208893
  • Cas Number : 54364-30-6
  • Molecular Weight : 183.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 253-254
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14531
Availability details for
Quantity
  • Availability
  • Add to Cart