ENA065086042 Enamine

6,8-dibromoquinazolin-4(3H)-one

  • MDL number MFCD00115129
  • Cas Number : 17518-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14352

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14352
Availability details for
Quantity
  • Availability
  • Add to Cart