ENA065086044 Enamine

2-(4-chlorophenyl)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD00141960
  • Cas Number : 54001-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213...215
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14870

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14870
Availability details for
Quantity
  • Availability
  • Add to Cart