ENA065086050 Enamine

5-[(2-phenylethyl)amino]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD03033838
  • Cas Number : 91129-85-0
  • Molecular Weight : 237.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189-190
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14876
Availability details for
Quantity
  • Availability
  • Add to Cart