ENA065086051 Enamine

(2,2,2-trifluoroethyl)urea

  • MDL number MFCD07346279
  • Cas Number : 819-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-110
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14541
Availability details for
Quantity
  • Availability
  • Add to Cart