ENA065086052 Enamine

2-methoxy-5-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD07343894
  • Cas Number : 81118-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-124
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14542

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-14542
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart