ENA065086067 Enamine

3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD05130922
  • Cas Number : 500025-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-106
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14359
Availability details for
Quantity
  • Availability
  • Add to Cart