ENA065086074 Enamine

4-(2-naphthyl)-4-oxobutanoic acid

  • MDL number MFCD00090143
  • Cas Number : 1590-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14900

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14900
Availability details for
Quantity
  • Availability
  • Add to Cart