ENA065086077 Enamine

4-{2-[(4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]acetamido}benzoic acid

  • MDL number MFCD00667233
  • Cas Number : 351006-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14903
Availability details for
Quantity
  • Availability
  • Add to Cart