ENA065086079 Enamine

2-[2-(thiophen-3-yl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD07339193
  • Cas Number : 863669-69-6
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14266
Availability details for
Quantity
  • Availability
  • Add to Cart