ENA065086087 Enamine

2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-4-methanesulfonylbutanoic acid

  • MDL number MFCD00414851
  • Cas Number : 355810-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14368
Availability details for
Quantity
  • Availability
  • Add to Cart