ENA065086093 Enamine

4-(carbamoylmethoxy)-3-methoxybenzoic acid

  • MDL number MFCD05039647
  • Cas Number : 869464-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14568
Availability details for
Quantity
  • Availability
  • Add to Cart