ENA065086096 Enamine

5-(3-bromophenyl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD05182411
  • Cas Number : 383130-99-2
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14922
Availability details for
Quantity
  • Availability
  • Add to Cart