ENA065086100 Enamine

4-[(2-nitrophenyl)amino]butanoic acid

  • MDL number MFCD07339190
  • Cas Number : 56636-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14272

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14272
Availability details for
Quantity
  • Availability
  • Add to Cart