ENA065086104 Enamine

3-amino-1-[(4-fluorophenyl)methyl]thiourea

  • MDL number MFCD00113788
  • Cas Number : 871548-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14930
Availability details for
Quantity
  • Availability
  • Add to Cart