ENA065086113 Enamine

2-amino-5-cyano-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD07339522
  • Cas Number : 138060-94-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-251
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14373
Availability details for
Quantity
  • Availability
  • Add to Cart