ENA065086115 Enamine

3-[3-(4-methylphenyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD06408967
  • Cas Number : 94192-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143...145
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14941

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14941
Availability details for
Quantity
  • Availability
  • Add to Cart