ENA065086117 Enamine

3-[3-(3,4-dimethoxyphenyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD05130953
  • Cas Number : 738593-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14575
Availability details for
Quantity
  • Availability
  • Add to Cart