ENA065086119 Enamine

5-(4-ethylphenyl)-2H-1,2,3,4-tetrazole

  • MDL number MFCD03114552
  • Cas Number : 460363-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14577
Availability details for
Quantity
  • Availability
  • Add to Cart