ENA065086141 Enamine

1-phenyl-4,5-dihydro-1H-1,2,3,4-tetrazol-5-one

  • MDL number MFCD01838361
  • Cas Number : 5097-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-189
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14590
Availability details for
Quantity
  • Availability
  • Add to Cart