ENA065086142 Enamine

ethyl (2Z)-4-chloro-3-hydroxy-2-(4-methyl-1,3-thiazol-2-yl)pent-2-enoate

  • MDL number MFCD07339186
  • Cas Number : 863669-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123...125
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14278

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14278
Availability details for
Quantity
  • Availability
  • Add to Cart