ENA065086143 Enamine

N-[3-(N'-hydroxycarbamimidoyl)phenyl]-2-(morpholin-4-yl)acetamide

  • Cas Number : 863669-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14279
Availability details for
Quantity
  • Availability
  • Add to Cart