ENA065086145 Enamine

4,6-dimethyl-2-(morpholine-4-carbonyl)thieno[2,3-b]pyridin-3-amine

  • MDL number MFCD01962581
  • Cas Number : 352024-83-0
  • Molecular Weight : 291.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-14971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14971
Availability details for
Quantity
  • Availability
  • Add to Cart