ENA065086149 Enamine

4-(3-fluorophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD03094297
  • Cas Number : 446065-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166-167
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-14975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14975
Availability details for
Quantity
  • Availability
  • Add to Cart