ENA065086150 Enamine

4-(1,3-benzothiazol-2-ylamino)butanoic acid

  • MDL number MFCD07339185
  • Cas Number : 104344-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14281

Safety Information

msds https://www.enaminestore.com/catalog/EN300-14281
Availability details for
Quantity
  • Availability
  • Add to Cart