ENA065086151 Enamine

5-chloro-2,3-dihydro-1-benzofuran-2-carboxylic acid

  • MDL number MFCD06753266
  • Cas Number : 34385-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-14386

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-14386
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart